5-Fluoro-2-nitrobenzaldehyde
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
006294-1G
006294-5G
006294-25G
FLUO006294-1GEA
0
GBP
FLUO006294-1G
FLUO006294-5G
FLUO006294-25G
5-Fluoro-2-nitrobenzaldehyde
5-Fluoro-2-nitrobenzaldehyde
Formula:
C₇H₄FNO₃ MW: 169.11 g/mol |
MDL Number:
MFCD00153175 CAS Number: 395-81-3 EINECS: 206-903-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...