4-Fluorobenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001460-100G 001460-500G
FLUO001460-100GEA 0 GBP
FLUO001460-100G FLUO001460-500G
4-Fluorobenzaldehyde
4-Fluorobenzaldehyde
Formula: C₇H₅FO
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: –10 °C
Density: 1.15 g/cm³ (20 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00003378
CAS Number: 459-57-4
EINECS: 207-293-6
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR