3-Fluorobenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001668-100G 001668-500G
FLUO001668-100GEA 0 GBP
FLUO001668-100G FLUO001668-500G
3-Fluorobenzaldehyde
3-Fluorobenzaldehyde
Formula: C₇H₅FO
Boiling Pt: 172…174 °C (1013 hPa)
Density: 1.7156 g/cm³ (20 °C)
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00003348
CAS Number: 456-48-4
EINECS: 207-266-9
UN: 1989
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR