3,5-Difluorobenzaldehyde
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001465-5G
001465-25G
001465-100G
FLUO001465-5GEA
0
GBP
FLUO001465-5G
FLUO001465-25G
FLUO001465-100G
3,5-Difluorobenzaldehyde
3,5-Difluorobenzaldehyde
Formula:
C₇H₄F₂O MW: 142.1 g/mol Boiling Pt: 61…63 °C (43 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00010329 CAS Number: 32085-88-4 UN: 1989 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...