2,5-Difluorobenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001932-25G 001932-100G
FLUO001932-25GEA 0 GBP
FLUO001932-25G FLUO001932-100G
2,5-Difluorobenzaldehyde
2,5-Difluorobenzaldehyde
Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 65…66 °C (20 mmHg)
Density: 1.3 g/cm³ (20 °C)
MDL Number: MFCD00010327
CAS Number: 2646-90-4
UN: 1989
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR