1,3-Difluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001232-100G 001232-500G 001232-2.5KG
FLUO001232-100GEA 0 GBP
FLUO001232-100G FLUO001232-500G FLUO001232-2.5KG
1,3-Difluorobenzene
1,3-Difluorobenzene
Formula: C₆H₄F₂
MW: 114.09 g/mol
Boiling Pt: 83 °C (1013 hPa)
Melting Pt: –59 °C
Density: 1.16 g/cm³ (20 °C)
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00000327
CAS Number: 372-18-9
EINECS: 206-746-5
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR