1,2-Difluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001169-100G
001169-500G
001169-2.5KG
FLUO001169-100GEA
0
GBP
FLUO001169-100G
FLUO001169-500G
FLUO001169-2.5KG
1,2-Difluorobenzene
1,2-Difluorobenzene
Formula:
C₆H₄F₂ MW: 114.09 g/mol Boiling Pt: 92 Melting Pt: –34 °C Density: 1.15 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000284 CAS Number: 367-11-3 EINECS: 206-680-7 UN: 1993 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...