1,2,4-Trifluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001792-25G
001792-100G
001792-500G
FLUO001792-25GEA
0
GBP
FLUO001792-25G
FLUO001792-100G
FLUO001792-500G
1,2,4-Trifluorobenzene
1,2,4-Trifluorobenzene
Formula:
C₆H₃F₃ Boiling Pt: 88 °C (1013 hPa) Melting Pt: –35 °C Density: 1.26 g/cm³ (20 °C) |
MDL Number:
MFCD00000305 CAS Number: 367-23-7 EINECS: 206-684-9 UN: 2387 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...