1,2,4,5-Tetrafluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
003312-100G
003312-10G
003312-50G
FLUO003312-100GEA
0
GBP
FLUO003312-100G
FLUO003312-10G
FLUO003312-50G
1,2,4,5-Tetrafluorobenzene
1,2,4,5-Tetrafluorobenzene
Formula:
C₆H₂F₄ MW: 150.08 g/mol Boiling Pt: 90 °C (1013 hPa) Melting Pt: 4 °C Density: 1.42 g/cm³ (20 °C) |
MDL Number:
MFCD00000307 CAS Number: 327-54-8 EINECS: 206-319-3 UN: 2387 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...