1,2,3-Trifluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001791-25G 001791-100G
FLUO001791-25GEA 0 GBP
FLUO001791-25G FLUO001791-100G
1,2,3-Trifluorobenzene
1,2,3-Trifluorobenzene
Formula: C₆H₃F₃
MW: 132.09 g/mol
Boiling Pt: 95 °C (1013 hPa)
Density: 1.28 g/cm³ (20 °C)
MDL Number: MFCD00012013
CAS Number: 1489-53-8
UN: 2387
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR