1,2,3,5-Tetrafluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003311-1G 003311-5G
FLUO003311-1GEA 0 GBP
FLUO003311-1G FLUO003311-5G
1,2,3,5-Tetrafluorobenzene
1,2,3,5-Tetrafluorobenzene
Formula: C₆H₂F₄
MW: 150.08 g/mol
Boiling Pt: 83 °C (1013 hPa)
Melting Pt: –48 °C
Density: 1.393 g/cm³ (20 °C)
MDL Number: MFCD00000302
CAS Number: 2367-82-0
EINECS: 219-126-4
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR