4-Chlorothiophenol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-Chloro-4-mercaptobenzene, 4-Chlorobenzene-1-thiol, 4-Chlorobenzenethiol, p-Chlorothiophenol
005205-100G
005205-500G
FLUO005205-100GEA
0
GBP
FLUO005205-100G
FLUO005205-500G
4-Chlorothiophenol
4-Chlorothiophenol
Formula:
C₆H₅ClS Boiling Pt: 205 °C (1013 hPa) Melting Pt: 49…53 °C Density: 1.224 g/cm³ (60 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004847 CAS Number: 106-54-7 EINECS: 203-408-9 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...