2,5-Dichlorothiophenol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
003354-5G
003354-25G
003354-100G
FLUO003354-5GEA
0
GBP
FLUO003354-5G
FLUO003354-25G
FLUO003354-100G
2,5-Dichlorothiophenol
2,5-Dichlorothiophenol
Formula:
C₆H₄Cl₂S Boiling Pt: 202 °C (1013 hPa) Melting Pt: 28 °C Density: 1.415 g/cm³ (30 °C) |
MDL Number:
MFCD00004831 CAS Number: 5858-18-4 EINECS: 227-486-9 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...