3,5-Dichlorothiophenol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
003357-25G
003357-100G
FLUO003357-25GEA
0
GBP
FLUO003357-25G
FLUO003357-100G
3,5-Dichlorothiophenol
3,5-Dichlorothiophenol
Formula:
C₆H₄Cl₂S Boiling Pt: 125…130 °C (12 mmHg) Melting Pt: 63.5 °C |
MDL Number:
MFCD00041423 CAS Number: 17231-94-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...