2,4-Dichlorothiophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,4-Dichlorobenzenethiol, 1,3-Dichloro-4-mercaptobenzene, 2,4-Dichlorobenzene-1-thiol

003353-5G 003353-100G 003353-25G
FLUO003353-5GEA 0 GBP
FLUO003353-5G FLUO003353-100G FLUO003353-25G
2,4-Dichlorothiophenol
2,4-Dichlorothiophenol
Formula: C₆H₄Cl₂S
Boiling Pt: 115 °C (8 mmHg)
Melting Pt: 22 °C
Density: 1,44 g/cm³ (20 °C)
MDL Number: MFCD00041421
CAS Number: 1122-41-4
EINECS: 214-346-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR