3,4-Dichlorothiophenol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
003356-5G
003356-25G
FLUO003356-5GEA
0
GBP
FLUO003356-5G
FLUO003356-25G
3,4-Dichlorothiophenol
3,4-Dichlorothiophenol
Formula:
C₆H₄Cl₂S Boiling Pt: 76…78 °C (0,3 mmHg) Melting Pt: 87…89 °C Density: 1,4175 g/cm³ (20 °C) |
MDL Number:
MFCD00004840 CAS Number: 5858-17-3 EINECS: 227-485-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...