2-(4-Chlorophenoxy)ethanol ≥98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Ethylene glycol mono(4-chlorophenyl) ether, 4-Chlorophenoxyethanol
A14508.06
A14508.14
A14508.06EA
20.1
GBP
A14508.06
A14508.14
2-(4-Chlorophenoxy)ethanol ≥98%
2-(4-Chlorophenoxy)ethanol
Purity: 98+%
CAS No.: 1892-43-9
Molecular Formula: C8H9ClO2
Formula Weight: 172.61
Refractive Index: 1.5510
Melting Point: 30°
Boiling Point: 135-136°/6mm
Beilstein Reference: 1365684
UN No.: 2811
MDL No.: MFCD00014036
Formula:
C₈H₉ClO₂ MW: 172.61 g/mol Boiling Pt: 135…136 °C (6 mmHg) Melting Pt: 30 °C Density: 1.2515 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00014036 CAS Number: 1892-43-9 EINECS: 217-578-7 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...