3-Chloro-2-nitroaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
011628-5G
011628-25G
FLUO011628-5GEA
0
GBP
FLUO011628-5G
FLUO011628-25G
3-Chloro-2-nitroaniline
3-Chloro-2-nitroaniline
Formula:
C₆H₅ClN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD01317803 CAS Number: 59483-54-4 EINECS: 261-782-9 UN: 2237 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...