3,5-Dichloroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

022133-1KG 022133-250G
FLUO022133-1KGEA 0 GBP
FLUO022133-1KG FLUO022133-250G
3,5-Dichloroaniline
3,5-Dichloroaniline
Formula: C₆H₅Cl₂N
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 51 °C
Density: 1.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007774
CAS Number: 626-43-7
EINECS: 210-948-9
UN: 3442
ADR: 6.1,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR