2,6-Dichloroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

005424-100G 005424-500G 005424-2.5KG
FLUO005424-100GEA 0 GBP
FLUO005424-100G FLUO005424-500G FLUO005424-2.5KG
2,6-Dichloroaniline
2,6-Dichloroaniline
Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 35…38 °C
Density: 1.526 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007675
CAS Number: 608-31-1
EINECS: 210-160-5
UN: 3442
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR