2,4-Dichloroaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
022088-1KG
022088-250G
FLUO022088-1KGEA
0
GBP
FLUO022088-1KG
FLUO022088-250G
2,4-Dichloroaniline
2,4-Dichloroaniline
Formula:
C₆H₅Cl₂N MW: 162.02 g/mol Boiling Pt: 245 °C (1013 hPa) Melting Pt: 59…62 °C Density: 1.567 g/cm³ (20 °C) Flash Pt: 149 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007661 CAS Number: 554-00-7 EINECS: 209-057-8 UN: 3442 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...