1-Chloro-4-iodobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001151-100G
001151-25G
FLUO001151-100GEA
0
GBP
FLUO001151-100G
FLUO001151-25G
1-Chloro-4-iodobenzene
1-Chloro-4-iodobenzene
Formula:
C₆H₄ClI Boiling Pt: 226 °C (1013 hPa) Melting Pt: 57 °C Density: 1,886 g/cm³ (57 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001053 CAS Number: 637-87-6 EINECS: 211-305-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...