1-Chloro-3-iodobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Chloroiodobenzene
001150-25G
001150-100G
FLUO001150-25GEA
0
GBP
FLUO001150-25G
FLUO001150-100G
1-Chloro-3-iodobenzene
1-Chloro-3-iodobenzene
Formula:
C₆H₄ClI MW: 238.45 g/mol Boiling Pt: 230 °C (1013 hPa) Melting Pt: 54,5 °C Density: 1,9255 g/cm³ (20 °C) |
MDL Number:
MFCD00001046 CAS Number: 625-99-0 EINECS: 210-920-6 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...