1-Chloro-2-iodobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Chloro-1-iodobenzene
001149-25G
001149-100G
FLUO001149-25GEA
0
GBP
FLUO001149-25G
FLUO001149-100G
1-Chloro-2-iodobenzene
1-Chloro-2-iodobenzene
Formula:
C₆H₄ClI MW: 238.45 g/mol Boiling Pt: 234 °C (1013 hPa) Melting Pt: 1 °C Density: 1,954 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001033 CAS Number: 615-41-8 EINECS: 210-423-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...