1,2-Dichloro-3-iodobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001165-25G
001165-100G
FLUO001165-25GEA
0
GBP
FLUO001165-25G
FLUO001165-100G
1,2-Dichloro-3-iodobenzene
Formula:
C₆H₃Cl₂I Boiling Pt: 242 °C (1013 hPa) Melting Pt: 35 °C |
MDL Number:
MFCD00001034 CAS Number: 2401-21-0 EINECS: 219-276-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...