3-Chloro-4-fluorophenol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
004458-100G
004458-25G
FLUO004458-100GEA
0
GBP
FLUO004458-100G
FLUO004458-25G
3-Chloro-4-fluorophenol
3-Chloro-4-fluorophenol
Formula:
C₆H₄ClFO MW: 146.55 g/mol Boiling Pt: 225 °C (1013 hPa) Melting Pt: 42…44 °C |
MDL Number:
MFCD00002257 CAS Number: 2613-23-2 EINECS: 220-042-5 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...