3-Chloro-1-fluoro-4-nitrobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-4-fluoro-1-nitrobenzene, 2-Chloro-4-fluoronitrobenzene

013801 013801-1G 013801-5G
FLUO013801EA 0 GBP
FLUO013801 FLUO013801-1G FLUO013801-5G
3-Chloro-1-fluoro-4-nitrobenzene
3-Chloro-1-fluoro-4-nitrobenzene
Formula: C₆H₃ClFNO₂
MW: 175.55 g/mol
MDL Number: MFCD03412200
CAS Number: 2106-50-5
EINECS: 218-286-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR