4-Chloro-2-fluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002196-25G 002196-100G 002196-10G
FLUO002196-25GEA 0 GBP
FLUO002196-25G FLUO002196-100G FLUO002196-10G
4-Chloro-2-fluoroaniline
4-Chloro-2-fluoroaniline
Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 104
Density: 1.299 g/cm³ (20 °C)
MDL Number: MFCD00010625
CAS Number: 57946-56-2
EINECS: 261-034-1
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR