3-Chloro-4-fluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001682-100G 001682-500G
FLUO001682-100GEA 0 GBP
FLUO001682-100G FLUO001682-500G
3-Chloro-4-fluoroaniline
3-Chloro-4-fluoroaniline
Formula: C₆H₅ClFN
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 41…44 °C
Density: 1.226 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007767
CAS Number: 367-21-5
EINECS: 206-682-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR