2-Chloro-6-fluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004417-1G 004417-5G 004417-25G
FLUO004417-1GEA 0 GBP
FLUO004417-1G FLUO004417-5G FLUO004417-25G
2-Chloro-6-fluoroaniline
2-Chloro-6-fluoroaniline
Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 183
Melting Pt: 31…32 °C
Density: 1.349 g/cm³ (20 °C)
MDL Number: MFCD00040309
CAS Number: 363-51-9
EINECS: 206-657-1
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR