2-Chloro-4-fluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001691-25G 001691-100G
FLUO001691-25GEA 0 GBP
FLUO001691-25G FLUO001691-100G
2-Chloro-4-fluoroaniline
2-Chloro-4-fluoroaniline
Formula: C₆H₅ClFN
Boiling Pt: 192 °C (1013 hPa)
Density: 1.21 g/cm³ (20 °C)
MDL Number: MFCD00042530
CAS Number: 2106-02-7
EINECS: 218-282-0
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR