4-Chloro-2-fluorobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
004430-25G
004430-100G
FLUO004430-25GEA
0
GBP
FLUO004430-25G
FLUO004430-100G
4-Chloro-2-fluorobenzoic acid
4-Chloro-2-fluorobenzoic acid
Formula:
C₇H₄ClFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00042468 CAS Number: 446-30-0 EINECS: 207-162-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...