4-Chloro-2,5-difluorobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001627-25G
001627-100G
FLUO001627-25GEA
0
GBP
FLUO001627-25G
FLUO001627-100G
4-Chloro-2,5-difluorobenzoic acid
4-Chloro-2,5-difluorobenzoic acid
Formula:
C₇H₃ClF₂O₂ MW: 192.55 g/mol Melting Pt: 153…158 °C |
MDL Number:
MFCD00077480 CAS Number: 132794-07-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...