2-Chloro-6-fluorobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001993-25G
001993-100G
001993-250G
FLUO001993-25GEA
0
GBP
FLUO001993-25G
FLUO001993-100G
FLUO001993-250G
2-Chloro-6-fluorobenzoic acid
2-Chloro-6-fluorobenzoic acid
Formula:
C₇H₄ClFO₂ MW: 174.56 g/mol Boiling Pt: 272 °C (1013 hPa) Melting Pt: 157…159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002417 CAS Number: 434-75-3 EINECS: 207-105-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...