4-Chloro-2-fluorobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009082-5G 009082-25G
FLUO009082-5GEA 0 GBP
FLUO009082-5G FLUO009082-25G
4-Chloro-2-fluorobenzonitrile
4-Chloro-2-fluorobenzonitrile
Formula: C₇H₃ClFN
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 57…58 °C
MDL Number: MFCD00143284
CAS Number: 57381-51-8
EINECS: 260-712-4
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR