5-Chloro-2-fluorobenzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005619-5G
005619-25G
FLUO005619-5GEA
0
GBP
FLUO005619-5G
FLUO005619-25G
5-Chloro-2-fluorobenzonitrile
5-Chloro-2-fluorobenzonitrile
Formula:
C₇H₃ClFN |
MDL Number:
MFCD01631434 CAS Number: 57381-34-7 EINECS: 260-710-3 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...