2-Chloro-6-fluorobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

005390-25G 005390-100G
FLUO005390-25GEA 0 GBP
FLUO005390-25G FLUO005390-100G
2-Chloro-6-fluorobenzonitrile
2-Chloro-6-fluorobenzonitrile
Formula: C₇H₃ClFN
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: 58…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00001780
CAS Number: 668-45-1
EINECS: 211-571-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR