2-Chloro-6-fluorobenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001426-100G 001426-500G
FLUO001426-100GEA 0 GBP
FLUO001426-100G FLUO001426-500G
2-Chloro-6-fluorobenzaldehyde
2-Chloro-6-fluorobenzaldehyde
Formula: C₇H₄ClFO
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: 36…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00003306
CAS Number: 387-45-1
EINECS: 206-860-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR