2-Chloro-4-fluorobenzaldehyde
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
FL4421.1
004421-5G
004421-25G
004421-100G
CHMPFL4421.1EA
0
GBP
CHMPFL4421.1
FLUO004421-5G
FLUO004421-25G
FLUO004421-100G
2-Chloro-4-fluorobenzaldehyde
2-Chloro-4-fluorobenzaldehyde
Formula:
C₇H₄ClFO Boiling Pt: 205 °C (1013 hPa) Melting Pt: 60 °C |
MDL Number:
MFCD00042527 CAS Number: 84194-36-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...