4-Chloro-1,2-difluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,4-Difluorochlorobenzene

002875-10G 002875-50G 002875-250G
FLUO002875-10GEA 0 GBP
FLUO002875-10G FLUO002875-50G FLUO002875-250G
4-Chloro-1,2-difluorobenzene
4-Chloro-1,2-difluorobenzene
Formula: C₆H₃ClF₂
Boiling Pt: 118…120 °C (1013 hPa)
Density: 1,35 g/cm³ (20 °C)
MDL Number: MFCD00042572
CAS Number: 696-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR