1-Chloro-2-fluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001145-100G
001145-500G
001145-25G
FLUO001145-100GEA
0
GBP
FLUO001145-100G
FLUO001145-500G
FLUO001145-25G
1-Chloro-2-fluorobenzene
1-Chloro-2-fluorobenzene
Formula:
C₆H₄ClF Boiling Pt: 137 °C (1013 hPa) Melting Pt: –42.5 °C Density: 1,244 g/cm³ (20 °C) |
MDL Number:
MFCD00000533 CAS Number: 348-51-6 EINECS: 206-476-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...