1,3-Dichloro-4-fluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,4-Dichlorofluorobenzene, 2,4-Dichloro-1-fluorobenzene

004707-100G 004707-500G
FLUO004707-100GEA 0 GBP
FLUO004707-100G FLUO004707-500G
1,3-Dichloro-4-fluorobenzene
1,3-Dichloro-4-fluorobenzene
Formula: C₆H₃Cl₂F
Boiling Pt: 172 °C (1013 hPa)
Melting Pt: –23 °C
Density: 1.4092 g/cm³ (20 °C)
MDL Number: MFCD00000580
CAS Number: 1435-48-9
EINECS: 406-160-1
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR