4-Chlorosalicylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

018377-10G 018377-25G
FLUO018377-10GEA 0 GBP
FLUO018377-10G FLUO018377-25G
4-Chlorosalicylic acid
4-Chlorosalicylic acid
Formula: C₇H₅ClO₃
MDL Number: MFCD00002449
CAS Number: 5106-98-9
EINECS: 225-829-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR