3-Chloro-2-nitrobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
023864-25G
023864-5G
FLUO023864-25GEA
0
GBP
FLUO023864-25G
FLUO023864-5G
3-Chloro-2-nitrobenzoic acid
3-Chloro-2-nitrobenzoic acid
Formula:
C₇H₄ClNO₄ Melting Pt: 236…240 °C Density: 1,566 g/cm³ (18 °C) |
MDL Number:
MFCD00007087 CAS Number: 4771-47-5 EINECS: 225-313-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...