4-Chloroanthranilic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Amino-4-chlorobenzoic acid
011358-250G
011358-100G
011358-25G
FLUO011358-250GEA
0
GBP
FLUO011358-250G
FLUO011358-100G
FLUO011358-25G
4-Chloroanthranilic acid
4-Chloroanthranilic acid
Formula:
C₇H₆ClNO₂ MW: 171.58 g/mol Boiling Pt: 330 °C (1013 hPa) Melting Pt: 240 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007778 CAS Number: 89-77-0 EINECS: 201-938-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...