2,6-Dichlorobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001380-25G
001380-100G
FLUO001380-25GEA
0
GBP
FLUO001380-25G
FLUO001380-100G
2,6-Dichlorobenzoic acid
2,6-Dichlorobenzoic acid
Formula:
C₇H₄Cl₂O₂ MW: 191.01 g/mol Boiling Pt: 299 Melting Pt: 138…139 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002418 CAS Number: 50-30-6 EINECS: 200-025-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...