2,3,5-Trichlorobenzoic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
002616-5G
002616-1G
FLUO002616-5GEA
0
GBP
FLUO002616-5G
FLUO002616-1G
2,3,5-Trichlorobenzoic acid
Formula:
C₇H₃Cl₃O₂ Boiling Pt: 324 °C (1013 hPa) Melting Pt: 171…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060674 CAS Number: 50-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...