4-Chloro-3,5-dinitrobenzonitrile ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

L03413.03 L03413.06
L03413.03EA 15.5 GBP
L03413.03 L03413.06
4-Chloro-3,5-dinitrobenzonitrile ≥97%
4-Chloro-3,5-dinitrobenzonitrile

Purity: 97% CAS No.: 1930-72-9 Molecular Formula: C7H2ClN3O4 Formula Weight: 227.56 Melting Point: 140-142° Beilstein Reference: 1990451 UN No.: 3439 MDL No.: MFCD00007067

Formula: C₇H₂ClN₃O₄
MDL Number: MFCD00007067
CAS Number: 1930-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR