2-Amino-6-chlorobenzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
018174-25G
018174-5G
FLUO018174-25GEA
0
GBP
FLUO018174-25G
FLUO018174-5G
2-Amino-6-chlorobenzonitrile
2-Amino-6-chlorobenzonitrile
Formula:
C₇H₅ClN₂ MW: 152.58 g/mol Boiling Pt: 301 °C (1013 hPa) Melting Pt: 136…138 °C |
MDL Number:
MFCD00015546 CAS Number: 6575-11-7 EINECS: 229-501-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...