2-Chloro-5-nitrobenzaldehyde
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
013138-25G
013138-100G
FLUO013138-25GEA
0
GBP
FLUO013138-25G
FLUO013138-100G
2-Chloro-5-nitrobenzaldehyde
2-Chloro-5-nitrobenzaldehyde
Formula:
C₇H₄ClNO₃ Melting Pt: 75…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007293 CAS Number: 6361-21-3 EINECS: 228-830-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...